Learn More
Medchemexpress LLC Glycocholic acid | 475-31-0 | 465.62 | 1 ML
SDP
Catalog No. 5000414851
Encompass_Preferred
Glycocholic acid is a bile acid with anticancer activity, targeting both pump resistance-related and non-pump resistance-related pathways. It increases the cytotoxicity of epirubicin and significantly increases its intracellular accumulation and absorption.
  • Intensifies epirubicin-induced apoptosis.
  • Reduces mRNA expression levels of human intestinal MDR1, MDR-associated protein (MRP)1, and MRP2.
  • Downregulates the MDR1 promoter region.
  • Suppresses the mRNA expression of Bcl-2 and induces the mRNA expression of Bax.
  • Increases the Bax-to-Bcl-2 ratio and the mRNA levels of p53, caspase-9, and -3.
  • Can control multidrug resistance (MDR) by modulating P-gp and MRPs, and regulating apoptosis-related pathways when combined with anticancer drugs.
Catalog No. 50-004-14851 Supplier Medchemexpress LLC Supplier No. HYN142310MM/1ML
Glycocholic acid is a bile acid with anticancer activity, targeting both pump resistance-related and non-pump resistance-related pathways. It increases the cytotoxicity of epirubicin and significantly increases its intracellular accumulation and absorption.
  • Intensifies epirubicin-induced apoptosis.
  • Reduces mRNA expression levels of human intestinal MDR1, MDR-associated protein (MRP)1, and MRP2.
  • Downregulates the MDR1 promoter region.
  • Suppresses the mRNA expression of Bcl-2 and induces the mRNA expression of Bax.
  • Increases the Bax-to-Bcl-2 ratio and the mRNA levels of p53, caspase-9, and -3.
  • Can control multidrug resistance (MDR) by modulating P-gp and MRPs, and regulating apoptosis-related pathways when combined with anticancer drugs.