Learn More
Medchemexpress LLC Glycocholic acid | 475-31-0 | 465.62 | 10 G
SDP
Catalog No. 5000415183
Encompass_Preferred
Glycocholic acid is a bile acid with anticancer activity, targeting against pump resistance-related and non-pump resistance-related pathways. It works by modulating P-gp and MRPs, and regulating apoptosis-related pathways when combined with anticancer drugs.
  • Increases cytotoxicity of epirubicin
  • Increases intracellular accumulation and absorption of epirubicin
  • Intensifies epirubicin-induced apoptosis
  • Reduces mRNA expression levels of human intestinal MDR1, MDR-associated protein (MRP)1, and MRP2
  • Downregulates the MDR1 promoter region
  • Suppresses mRNA expression of Bcl-2
  • Induces mRNA expression of Bax
  • Increases Bax-to-Bcl-2 ratio and mRNA levels of p53, caspase-9 and -3
Catalog No. 50-004-15183 Supplier Medchemexpress LLC Supplier No. HYN142310G
Glycocholic acid is a bile acid with anticancer activity, targeting against pump resistance-related and non-pump resistance-related pathways. It works by modulating P-gp and MRPs, and regulating apoptosis-related pathways when combined with anticancer drugs.
  • Increases cytotoxicity of epirubicin
  • Increases intracellular accumulation and absorption of epirubicin
  • Intensifies epirubicin-induced apoptosis
  • Reduces mRNA expression levels of human intestinal MDR1, MDR-associated protein (MRP)1, and MRP2
  • Downregulates the MDR1 promoter region
  • Suppresses mRNA expression of Bcl-2
  • Induces mRNA expression of Bax
  • Increases Bax-to-Bcl-2 ratio and mRNA levels of p53, caspase-9 and -3