Learn More
Medchemexpress LLC Glycocholic acid-d4 | 1201918-15-1 | 469.65 | 1 MG
SDP
Catalog No. 5000414535
Encompass_Preferred
Glycocholic acid-d4 is the deuterium-labeled Glycocholic acid. Glycocholic acid is a bile acid that exhibits anticancer activity by targeting pump resistance-related and non-pump resistance-related pathways.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14535 Supplier Medchemexpress LLC Supplier No. HYN1423S1MG
Glycocholic acid-d4 is the deuterium-labeled Glycocholic acid. Glycocholic acid is a bile acid that exhibits anticancer activity by targeting pump resistance-related and non-pump resistance-related pathways.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.