Learn More
Cayman Chemical Glycocholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1201918-15-1, 1 mg
SDP
Catalog No. 502985768
Encompass_Preferred
An internal standard for the quantification of glycocholic acid by GC- or LC-MS
Catalog No. 50-298-5768 Supplier Cayman Chemical Supplier No. 218891 MG
An internal standard for the quantification of glycocholic acid by GC- or LC-MS