Learn More
Cayman Chemical GlycocholIc AcId-d4 5mg
SDP
Catalog No. 502987993
Encompass_Preferred
An internal standard for the quantification of glycocholic acid by GC- or LC-MS
This item is not returnable. View return policy
Catalog No. 50-298-7993 Supplier Cayman Chemical Supplier No. 218895 MG
An internal standard for the quantification of glycocholic acid by GC- or LC-MS