Learn More
Medchemexpress LLC Glycocholic acid-d5 | 2170091-95-7 | 470.65 | 1 MG
SDP
Catalog No. 5000414652
Encompass_Preferred
Glycocholic acid-d5 is the deuterium labeled Glycocholic acid, which is a bile acid exhibiting anticancer activity by targeting both pump resistance-related and non-pump resistance-related pathways. This compound is intended for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-14652 Supplier Medchemexpress LLC Supplier No. HYN1423S11MG
Glycocholic acid-d5 is the deuterium labeled Glycocholic acid, which is a bile acid exhibiting anticancer activity by targeting both pump resistance-related and non-pump resistance-related pathways. This compound is intended for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.