Learn More
Medchemexpress LLC Glycocholic acid (Standard) | 475-31-0 | 465.62 | 100 MG
SDP
Catalog No. 5000416268
Encompass_Preferred
Glycocholic acid (Standard) is the analytical standard of Glycocholic acid, intended for research and analytical applications. It is a bile acid with anticancer activity, targeting against pump resistance-related and non-pump resistance-related pathways.
  • Analytical standard for Glycocholic acid.
  • Intended for research and analytical applications.
  • Has anticancer activity.
  • Targets pump resistance-related and non-pump resistance-related pathways.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-16268 Supplier Medchemexpress LLC Supplier No. HYN1423R100MG
Add to Cart
Add to Cart
Glycocholic acid (Standard) is the analytical standard of Glycocholic acid, intended for research and analytical applications. It is a bile acid with anticancer activity, targeting against pump resistance-related and non-pump resistance-related pathways.
  • Analytical standard for Glycocholic acid.
  • Intended for research and analytical applications.
  • Has anticancer activity.
  • Targets pump resistance-related and non-pump resistance-related pathways.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.