Learn More
Medchemexpress LLC Glycocholic acid (Standard) | 475-31-0 | 465.62 | 5 MG
SDP
Catalog No. 5000415322
Encompass_Preferred
Glycocholic acid (Standard) is the analytical standard of Glycocholic acid, intended for research and analytical applications. It is a bile acid with anticancer activity, targeting against pump resistance-related and non-pump resistance-related pathways. This compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications.
  • Exhibits anticancer activity.
  • Targets pump resistance-related and non-pump resistance-related pathways.
  • Suitable for HPLC, GC, and MS experiments.
  • Purity of 99.90%.
  • Appears as a white to off-white solid.
Catalog No. 50-004-15322 Supplier Medchemexpress LLC Supplier No. HYN1423R5MG
Add to Cart
Add to Cart
Glycocholic acid (Standard) is the analytical standard of Glycocholic acid, intended for research and analytical applications. It is a bile acid with anticancer activity, targeting against pump resistance-related and non-pump resistance-related pathways. This compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications.
  • Exhibits anticancer activity.
  • Targets pump resistance-related and non-pump resistance-related pathways.
  • Suitable for HPLC, GC, and MS experiments.
  • Purity of 99.90%.
  • Appears as a white to off-white solid.