Learn More
Medchemexpress LLC Glycocholic acid (Standard) | 475-31-0 | 465.62 | 50 MG
SDP
Catalog No. 5000416106
Encompass_Preferred
Glycocholic acid (Standard) is an analytical standard of Glycocholic acid, intended for research and analytical applications. This bile acid exhibits anticancer activity by targeting pump resistance-related and non-pump resistance-related pathways. The compound serves as a reference standard in assays and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable for HPLC, GC, and MS
  • Exhibits anticancer activity
Catalog No. 50-004-16106 Supplier Medchemexpress LLC Supplier No. HYN1423R50MG
Add to Cart
Add to Cart
Glycocholic acid (Standard) is an analytical standard of Glycocholic acid, intended for research and analytical applications. This bile acid exhibits anticancer activity by targeting pump resistance-related and non-pump resistance-related pathways. The compound serves as a reference standard in assays and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable for HPLC, GC, and MS
  • Exhibits anticancer activity