Learn More
Medchemexpress LLC Glycocyamine-15N,13C2 | 2483829-93-0 | 97.4% | 120.09 | C13C2H7N215NO2 | 10 MG
SDP
Catalog No. 5000299509
Encompass_Preferred
Glycocyamine-15N,13C2 is a 13C- and 15N-labeled form of glycocyamine (guanidinoacetic acid) supplied as a stable isotope-labeled powder for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is provided in small-mass sizes suitable for analytical research, calibration, and method development.
  • Isotopically labeled with 13C and 15N for accurate tracing and quantitation.
  • High purity suitable for analytical applications.
  • Available in small sizes for standard preparation and calibration.
  • Stable solid form with specified storage conditions for long-term use.
  • Compatible with NMR, GC-MS, and LC-MS workflows as an internal standard.
Catalog No. 50-002-99509 Supplier Medchemexpress LLC Supplier No. HYW021448S110MG
Glycocyamine-15N,13C2 is a 13C- and 15N-labeled form of glycocyamine (guanidinoacetic acid) supplied as a stable isotope-labeled powder for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is provided in small-mass sizes suitable for analytical research, calibration, and method development.
  • Isotopically labeled with 13C and 15N for accurate tracing and quantitation.
  • High purity suitable for analytical applications.
  • Available in small sizes for standard preparation and calibration.
  • Stable solid form with specified storage conditions for long-term use.
  • Compatible with NMR, GC-MS, and LC-MS workflows as an internal standard.