Learn More
Medchemexpress LLC Glycocyamine-15N,13C2 | 2483829-93-0 | 97.4% | 120.09 | C13C2H7N215NO2 | 5 MG
SDP
Catalog No. 5000300490
Encompass_Preferred
Glycocyamine-15N,13C2 is an isotopically labeled form of glycocyamine (guanidinoacetic acid) intended for use as a stable-isotope tracer and reference standard in analytical and biochemical research. Supplied as a powder for preparation of standard solutions and experimental assays; for research use only.
  • Isotopically labeled with 15N and 13C for tracer and quantification studies.
  • High purity (97.4%) suitable for analytical applications.
  • Supplied as a powder for easy dissolution and preparation of standards.
  • Compatible with mass spectrometry and NMR workflows.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-003-00490 Supplier Medchemexpress LLC Supplier No. HYW021448S15MG
Glycocyamine-15N,13C2 is an isotopically labeled form of glycocyamine (guanidinoacetic acid) intended for use as a stable-isotope tracer and reference standard in analytical and biochemical research. Supplied as a powder for preparation of standard solutions and experimental assays; for research use only.
  • Isotopically labeled with 15N and 13C for tracer and quantification studies.
  • High purity (97.4%) suitable for analytical applications.
  • Supplied as a powder for easy dissolution and preparation of standards.
  • Compatible with mass spectrometry and NMR workflows.
  • Stable when stored under recommended conditions for long-term use.