Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycocyamine-d2 (guanidinoacetic acid-d2) | 1173020-63-7 | 99.9% | 119.12 | C3H5D2N3O2 | 1 MG
SDP

Catalog No. 5000301997
Encompass_Preferred

Glycocyamine-d2 is the deuterium-labeled analog of glycocyamine (guanidinoacetic acid), a metabolic precursor of creatine. It is provided as a stable-isotope standard for research and analytical applications, including quantitative mass spectrometry and metabolic studies.

  • Stable-isotope labeled deuterated analog for tracer and internal standard use.
  • High purity (99.9%) suitable for analytical workflows.
  • Molecular formula C3H5D2N3O2 and molecular weight 119.12.
  • Available in small to bulk quantities (1-500 mg) for method development and routine assays.
  • Suitable for use in mass spectrometry and metabolic investigations.

Catalog No. 50-003-01997 Supplier Medchemexpress LLC Supplier No. HYW021448S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycocyamine-d2 is the deuterium-labeled analog of glycocyamine (guanidinoacetic acid), a metabolic precursor of creatine. It is provided as a stable-isotope standard for research and analytical applications, including quantitative mass spectrometry and metabolic studies.

  • Stable-isotope labeled deuterated analog for tracer and internal standard use.
  • High purity (99.9%) suitable for analytical workflows.
  • Molecular formula C3H5D2N3O2 and molecular weight 119.12.
  • Available in small to bulk quantities (1-500 mg) for method development and routine assays.
  • Suitable for use in mass spectrometry and metabolic investigations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.