Learn More
Cayman Chemical GlycodeoxycholIc ACD MaxSpec
SDP
Catalog No. 502988093
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-298-8093 Supplier Cayman Chemical Supplier No. 31599100 UG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications