Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycodeoxycholic acid-d4 | 1069132-37-1 | 99.9% | 453.65 | C26H39D4NO5 | 5MG
SDP

Catalog No. 5000236493
Encompass_Preferred

Glycodeoxycholic acid-d4 is a deuterium-labeled stable isotope of glycodeoxycholic acid supplied as a white to off-white solid for research use. It serves as a tracer or internal standard in quantitative analytical methods such as LC-MS, GC-MS, and NMR for accurate measurement of glycodeoxycholic acid in biological samples.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High chemical purity: 99.9%.
  • CAS number 1069132-37-1 for reference.
  • Chemical formula C26H39D4NO5 and molecular weight 453.65 g/mol.
  • Available in small pack sizes suitable for standards (1 mg to 50 mg).
  • White to off-white solid appearance.
  • Store at -20°C and protect from light; in solvent, store at -80°C for long-term stability.

Catalog No. 50-002-36493 Supplier Medchemexpress LLC Supplier No. HY125731S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycodeoxycholic acid-d4 is a deuterium-labeled stable isotope of glycodeoxycholic acid supplied as a white to off-white solid for research use. It serves as a tracer or internal standard in quantitative analytical methods such as LC-MS, GC-MS, and NMR for accurate measurement of glycodeoxycholic acid in biological samples.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High chemical purity: 99.9%.
  • CAS number 1069132-37-1 for reference.
  • Chemical formula C26H39D4NO5 and molecular weight 453.65 g/mol.
  • Available in small pack sizes suitable for standards (1 mg to 50 mg).
  • White to off-white solid appearance.
  • Store at -20°C and protect from light; in solvent, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.