Learn More
Cayman Chemical GlycodeoxycholIc AcId-d4 500ug
SDP
Catalog No. 502990940
Encompass_Preferred
An internal standard for the quantification of glycodeoxycholic acid by GC- or LC-MS
Catalog No. 50-299-0940 Supplier Cayman Chemical Supplier No. 31310500 UG
An internal standard for the quantification of glycodeoxycholic acid by GC- or LC-MS