Learn More
Cayman Chemical Glycodeoxycholic Acid (hydrate), Purity: ≥95%, CAS: 1079043-81-4, 1 g
SDP
Catalog No. 502983766
Encompass_Preferred
A glycine-conjugated form of deoxycholic acid; induces a reversible, concentration-dependent reduction in myogenic tone in rats and decreases expression of the gene encoding CYP7A1 in rabbits; serum levels are elevated in non-surviving patients with AALF compared with survivors; plasma levels are increased in the plasma of patients with asthma
Catalog No. 50-298-3766 Supplier Cayman Chemical Supplier No. 202741 G
A glycine-conjugated form of deoxycholic acid; induces a reversible, concentration-dependent reduction in myogenic tone in rats and decreases expression of the gene encoding CYP7A1 in rabbits; serum levels are elevated in non-surviving patients with AALF compared with survivors; plasma levels are increased in the plasma of patients with asthma