Learn More
Medchemexpress LLC Glycogen, from bovine liver | 9005-79-2 | ≥85% | 100 MG
SDP
Catalog No. 5000436378
Encompass_Preferred
Glycogen, from bovine liver, ≥85% is a branched polymer of glucose synthesized by animal cells for energy storage and release that can be isolated from bovine liver. It is a biochemical reagent that can be used for life science related research.
  • Can be used as a standard or substrate in assays to detect and quantify liver glycogen content.
  • Can be used for carbohydrate storage and metabolism research.
  • Used to study various enzymes such as alpha-glucosidase(s) and glycogen phosphorylase(s).
  • Can serve as a substrate to identify and characterize its metabolizing enzymes.
Catalog No. 50-004-36378 Supplier Medchemexpress LLC Supplier No. HY113511C100MG
Add to Cart
Add to Cart
Glycogen, from bovine liver, ≥85% is a branched polymer of glucose synthesized by animal cells for energy storage and release that can be isolated from bovine liver. It is a biochemical reagent that can be used for life science related research.
  • Can be used as a standard or substrate in assays to detect and quantify liver glycogen content.
  • Can be used for carbohydrate storage and metabolism research.
  • Used to study various enzymes such as alpha-glucosidase(s) and glycogen phosphorylase(s).
  • Can serve as a substrate to identify and characterize its metabolizing enzymes.