Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycogen from Mytilus edul5G
Catalog No. 502902576
Encompass
Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly alpha1-4 glycosidic bonds with branches created through alpha1-6 glycosidic bonds.
Catalog No. 50-290-2576 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G15085G
Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly alpha1-4 glycosidic bonds with branches created through alpha1-6 glycosidic bonds.