Learn More
Crescent Chemical Co Inc Glycogen from Oyster, 1mL
SDP
Supplier:  Crescent Chemical Co Inc SE39766.01
Encompass_Preferred
DNase/RNase not detected. 20 mg/ml solution in redistilled water. Suitable as a carrier molecule for DNA and RNA, replacing tRNA and sonicated DNA. Storage Conditions: -15 °C TO -25 °C
Catalog No. 50-248-451