Learn More
Crescent Chemical Co Inc GLYCOGEN FROM OYSTER
SDP
Catalog No. 50247691
Encompass_Preferred
Glycogen from oyster research grade Substrate for glycogenphosphorylase (EC 2.4.1.1). Suitable as a carrier molecule for DNA and RNA in precipitation reactions, replacing tRNA and sonicated DN Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number
Catalog No. 50-247-691 Supplier Crescent Chemical Co Inc Supplier No. SE23550.02
Glycogen from oyster research grade Substrate for glycogenphosphorylase (EC 2.4.1.1). Suitable as a carrier molecule for DNA and RNA in precipitation reactions, replacing tRNA and sonicated DN Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number