Learn More
Medchemexpress LLC Glycogen (from rabbit liver) | 9005-79-2 | MFCD00081547 | 91.0% | Not specified (polymeric, variable molecular weight) | (C6H10O5)n | 5 MG
SDP
Catalog No. 5000264816
Encompass_Preferred
Glycogen from rabbit liver is a highly branched glucose polysaccharide provided as a solid for research use. It functions as the animal storage form of glucose and is used as a reagent, standard, or carrier in various biochemical and molecular biology assays. Certificate of analysis and safety data are available to confirm lot-specific properties.
  • High branching and biological relevance for metabolism studies.
  • Suitable as a reagent or standard in enzymatic and biochemical assays.
  • Provides a reliable carrier for nucleic acid precipitation and detection assays.
  • Supplied as a stable powdered solid for ease of handling and storage.
  • Lot-specific purity and analytical data available via certificate of analysis.
Catalog No. 50-002-64816 Supplier Medchemexpress LLC Supplier No. HY113511B5MG
Glycogen from rabbit liver is a highly branched glucose polysaccharide provided as a solid for research use. It functions as the animal storage form of glucose and is used as a reagent, standard, or carrier in various biochemical and molecular biology assays. Certificate of analysis and safety data are available to confirm lot-specific properties.
  • High branching and biological relevance for metabolism studies.
  • Suitable as a reagent or standard in enzymatic and biochemical assays.
  • Provides a reliable carrier for nucleic acid precipitation and detection assays.
  • Supplied as a stable powdered solid for ease of handling and storage.
  • Lot-specific purity and analytical data available via certificate of analysis.