Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycogen from rabbit liver1G
Catalog No. 502902025
Encompass
Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly alpha1-4 glycosidic bonds with branches created through alpha1-6 glycosidic bonds.
Catalog No. 50-290-2025 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G88761G
Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly alpha1-4 glycosidic bonds with branches created through alpha1-6 glycosidic bonds.