Learn More
Medchemexpress LLC Glycogen, mussel | 9005-79-2 | 99.3% | 1 G
SDP
Catalog No. 5000437491
Encompass_Preferred
Glycogen, Mussel is a glycolytic intermediate and high-energy phosphate that can serve as a form of energy storage in humans, animals, fungi, and bacteria. It is a branched polymer of glucose primarily stored in the liver and skeletal muscle in the human body, providing glucose to the bloodstream during fasting and to muscle cells during contraction.
  • Functions as a glycolytic intermediate and high-energy phosphate.
  • Serves as an energy storage form in various organisms, including humans, animals, fungi, and bacteria.
  • A branched polymer of glucose.
  • Supplies glucose to the bloodstream during fasting periods.
  • Provides glucose to muscle cells during muscle contraction.
  • For research use only.
Catalog No. 50-004-37491 Supplier Medchemexpress LLC Supplier No. HY1135111G
Glycogen, Mussel is a glycolytic intermediate and high-energy phosphate that can serve as a form of energy storage in humans, animals, fungi, and bacteria. It is a branched polymer of glucose primarily stored in the liver and skeletal muscle in the human body, providing glucose to the bloodstream during fasting and to muscle cells during contraction.
  • Functions as a glycolytic intermediate and high-energy phosphate.
  • Serves as an energy storage form in various organisms, including humans, animals, fungi, and bacteria.
  • A branched polymer of glucose.
  • Supplies glucose to the bloodstream during fasting periods.
  • Provides glucose to muscle cells during muscle contraction.
  • For research use only.