Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycogen, Mussel | 9005-79-2 | 99.3% | 50 MG
SDP

Catalog No. 5000436520
Encompass_Preferred

Glycogen, Mussel is a glycolytic intermediate and high-energy phosphate that serves as a form of energy storage in humans, animals, fungi, and bacteria. It is a branched polymer of glucose stored mainly in the liver and skeletal muscle in the human body that supplies glucose to the bloodstream during fasting periods and to muscle cells during muscle contraction.

  • Glycolytic intermediate
  • High-energy phosphate
  • Serves as a form of energy storage
  • Endogenous metabolite
  • Classified as a saccharide
  • Appearance is solid
  • Color is white to off-white
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-36520 Supplier Medchemexpress LLC Supplier No. HY11351150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycogen, Mussel is a glycolytic intermediate and high-energy phosphate that serves as a form of energy storage in humans, animals, fungi, and bacteria. It is a branched polymer of glucose stored mainly in the liver and skeletal muscle in the human body that supplies glucose to the bloodstream during fasting periods and to muscle cells during muscle contraction.

  • Glycolytic intermediate
  • High-energy phosphate
  • Serves as a form of energy storage
  • Endogenous metabolite
  • Classified as a saccharide
  • Appearance is solid
  • Color is white to off-white
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.