Learn More
Medchemexpress LLC Glycogen, Oysters | 9005-79-2 | 500 MG
SDP
Catalog No. 5000435904
Encompass_Preferred
Glycogen, Oysters is a polysaccharide extracted from oysters and serves as the storage form of glucose in oysters. It belongs to the intermediate products of glycolysis and high-energy phosphates, and is also an endogenous metabolite.
  • It is a polysaccharide extracted from oysters.
  • It serves as the storage form of glucose in oysters.
  • It belongs to the intermediate products of glycolysis and high-energy phosphates.
  • It is an endogenous metabolite.
  • It has a highly multi-branched spherical structure.
  • It can be digested by salivary and pancreatic α-amylase to produce various branched oligosaccharides.
Catalog No. 50-004-35904 Supplier Medchemexpress LLC Supplier No. HY113511A500MG
Glycogen, Oysters is a polysaccharide extracted from oysters and serves as the storage form of glucose in oysters. It belongs to the intermediate products of glycolysis and high-energy phosphates, and is also an endogenous metabolite.
  • It is a polysaccharide extracted from oysters.
  • It serves as the storage form of glucose in oysters.
  • It belongs to the intermediate products of glycolysis and high-energy phosphates.
  • It is an endogenous metabolite.
  • It has a highly multi-branched spherical structure.
  • It can be digested by salivary and pancreatic α-amylase to produce various branched oligosaccharides.