Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycogen from oysters, MP Biomedicals™

Catalog No. MP1GLYCO001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1GLYCO001 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1GLYCO001 Supplier MP Biomedicals, Inc Supplier No. 11GLYCO001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycogen from oysters, Molecular Biology Grade

  • Glycogen is used as a carrier for the precipitation of nucleic acids (DNA or RNA).
  • Glycogen is an inert material and does not interfere with molecular biology experiments.
  • It can replace tRNAs or sonicated DNAs as an additive for the precipitation of nucleic acids.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μL
Grade Molecular Biology
Physical Form Solution
storageNote1 -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.