Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycogen, from Oysters, Ultrapure

Catalog No. AAJ1644514 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1644514 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ1644514 Supplier Thermo Scientific Supplier No. J1644514

missing translation for 'validMainframeMsg'

CAS: 9005-79-2 | C24H42O21 | 666.579 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-79-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00081547
InChI Key BYSGBSNPRWKUQH-UJDJLXLFSA-N
Synonym glycogen, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2 r,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2 s,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2 s,3 r,4 s,5 s,6 r-2-2 r,3 s,4 r,5 r,6 r-4,5-dihydroxy-3-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2 r,3 s,4 r,5 r,6 s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439177
ChEBI CHEBI:28087
IUPAC Name (2 S,3 R,4 S,5 S,6 R)-2-[[(2 R,3 S,4 R,5 R,6 R)-4,5-dihydroxy-3-[(2 R,3 R,4 S,5 S,6 R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2 R,3 S,4 R,5 R,6 S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O

Specifications

Packaging Plastic bottle
Quantity 25 g
Grade Ultrapure
Physical Form Powder
Chemical Name or Material Glycogen
Hazard Category H500
EINECSNumber 232-683-8
RTECSNumber MC2700000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.