Learn More
Cayman Chemical Glycogen phsphoylase InHBI 1mg
SDP
Catalog No. 502965053
Encompass_Preferred
A cell-permeable acyl urea first identified as an inhibitor of human liver glycogen phosphorylase (IC50 = 53 nM); blocks glucagon-induced hepatic glycogenolysis in vivo
Catalog No. 50-296-5053 Supplier Cayman Chemical Supplier No. 175781 MG
A cell-permeable acyl urea first identified as an inhibitor of human liver glycogen phosphorylase (IC50 = 53 nM); blocks glucagon-induced hepatic glycogenolysis in vivo