Learn More
Cayman Chemical Glycogen Phosphorylase Inhibitor, Purity: ≥98%, CAS: 648926-15-2, 5 mg
SDP
Catalog No. 502967358
Encompass_Preferred
A cell-permeable acyl urea first identified as an inhibitor of human liver glycogen phosphorylase (IC50 = 53 nM); blocks glucagon-induced hepatic glycogenolysis in vivo
Catalog No. 50-296-7358 Supplier Cayman Chemical Supplier No. 175785 MG
A cell-permeable acyl urea first identified as an inhibitor of human liver glycogen phosphorylase (IC50 = 53 nM); blocks glucagon-induced hepatic glycogenolysis in vivo