Learn More
Medchemexpress LLC Glycogen, from rabbit liver | 9005-79-2 | 91.0% | 50 MG
SDP
Catalog No. 5000437469
Encompass_Preferred
Glycogen, from rabbit liver, ≥85% is glycogen extracted from rabbit liver. It is a branched polysaccharide condensed from glucose and serves as a storage form of glucose in the body. It plays an important role in physiological processes such as maintaining blood sugar and providing energy.
  • For research use only
Catalog No. 50-004-37469 Supplier Medchemexpress LLC Supplier No. HY113511B50MG
Add to Cart
Add to Cart
Glycogen, from rabbit liver, ≥85% is glycogen extracted from rabbit liver. It is a branched polysaccharide condensed from glucose and serves as a storage form of glucose in the body. It plays an important role in physiological processes such as maintaining blood sugar and providing energy.
  • For research use only