Learn More
Medchemexpress LLC Glycogen synthase 1/GYS1 Antibody (YA400) | Protein A affinity purified | 81 kDa | 10 UL
SDP
Catalog No. 5000397796
Encompass_Preferred
Glycogen synthase 1/GYS1 Antibody (YA400) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Glycogen synthase 1 (GYS1). GYS1 plays a central role in regulating glycogen levels within cells, contributing to energy storage and metabolism by sequentially adding glucose residues to growing glycogen chains during synthesis.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets glycogen synthase 1 (GYS1)
  • Crucial for glycogen synthesis
  • Regulates cellular glycogen levels
  • Contributes to energy storage and metabolism
Catalog No. 50-003-97796 Supplier Medchemexpress LLC Supplier No. HYP8014510UL
Glycogen synthase 1/GYS1 Antibody (YA400) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Glycogen synthase 1 (GYS1). GYS1 plays a central role in regulating glycogen levels within cells, contributing to energy storage and metabolism by sequentially adding glucose residues to growing glycogen chains during synthesis.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets glycogen synthase 1 (GYS1)
  • Crucial for glycogen synthesis
  • Regulates cellular glycogen levels
  • Contributes to energy storage and metabolism