Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycogen synthase 1/GYS1 Antibody (YA400) | Protein A affinity purified | 81 kDa | 50 UL
SDP

Catalog No. 5000396864
Encompass_Preferred

Glycogen synthase 1/GYS1 Antibody (YA400) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Glycogen synthase 1/GYS1. This enzyme plays a central role in regulating glycogen levels within cells by transferring a glycosyl residue from UDP-Glc to alpha-1,4-glucan, crucial for glycogen synthesis. It is protein A affinity purified and reactive with human and mouse samples.

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Conjugation: Non-conjugated
  • Molecular weight: 81 kDa
  • Purity: Protein A affinity purified
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide
  • Applications: WB, ICC/IF, IHC-P, FC, IP
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-003-96864 Supplier Medchemexpress LLC Supplier No. HYP8014550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycogen synthase 1/GYS1 Antibody (YA400) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Glycogen synthase 1/GYS1. This enzyme plays a central role in regulating glycogen levels within cells by transferring a glycosyl residue from UDP-Glc to alpha-1,4-glucan, crucial for glycogen synthesis. It is protein A affinity purified and reactive with human and mouse samples.

  • Host: Rabbit
  • Reactivity: Human, Mouse
  • Conjugation: Non-conjugated
  • Molecular weight: 81 kDa
  • Purity: Protein A affinity purified
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide
  • Applications: WB, ICC/IF, IHC-P, FC, IP
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.