Learn More
Cell Signaling Technology Glycogen Synthase 2 Monoclonal Antibody, Rabbit, Human, Mouse, Rat, 100 μL
SDP
Catalog No. 502731859
Encompass
Glycogen Synthase 2 (GYS2) (E5E4V) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total GYS2 protein. This antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human GYS2 protein. It is suitable for applications including Western blotting and immunoprecipitation and does not cross-react with GYS1.
  • Validated applications include Western blotting and immunoprecipitation.
  • Specific to endogenous levels of total protein.
  • Displays no cross-reactivity with the GYS1 isoform.
  • Species reactivity includes human, mouse, and rat.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
Catalog No. 50-273-1859 Supplier Cell Signaling Technology Supplier No. 71217S
Glycogen Synthase 2 (GYS2) (E5E4V) Rabbit Monoclonal Antibody is designed to detect endogenous levels of total GYS2 protein. This antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human GYS2 protein. It is suitable for applications including Western blotting and immunoprecipitation and does not cross-react with GYS1.
  • Validated applications include Western blotting and immunoprecipitation.
  • Specific to endogenous levels of total protein.
  • Displays no cross-reactivity with the GYS1 isoform.
  • Species reactivity includes human, mouse, and rat.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.