Learn More
Cayman Chemical GlycohyocholIc AcId 10mg
SDP
Catalog No. 502995374
Encompass_Preferred
A glycine-conjugated form of hyocholic acid; upregulated 6.6-fold in the serum of patients with cirrhosis induced by HCV compared to healthy controls; plasma levels increase in patients who are no longer diabetic following gastric bypass surgery
Catalog No. 50-299-5374 Supplier Cayman Chemical Supplier No. 2267010 MG
A glycine-conjugated form of hyocholic acid; upregulated 6.6-fold in the serum of patients with cirrhosis induced by HCV compared to healthy controls; plasma levels increase in patients who are no longer diabetic following gastric bypass surgery