Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycohyocholic acid | 32747-08-3 | >98.0% | 50 MG
SDP

Catalog No. 5000231350
Encompass_Preferred

Analytical reference standard of glycohyocholic acid, a glycine-conjugated bile acid intended for research and analytical applications. Typical pack size is 50 MG; chemical formula C26H43NO6 and molecular weight ~465.6 g/mol. Purity is reported as ≥98% by some suppliers; verify the certificate of analysis.

  • Analytical reference standard for use in research and analytical assays.
  • Glycine-conjugated bile acid structure, useful as a metabolite standard.
  • Chemical formula: C26H43NO6.
  • Molecular weight: ~465.6 g/mol.
  • Supplied as a 50 MG solid suitable for analytical preparation.
  • Purity reported as ≥98% by some suppliers; check the certificate of analysis.

Catalog No. 50-002-31350 Supplier Medchemexpress LLC Supplier No. HY12214150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Analytical reference standard of glycohyocholic acid, a glycine-conjugated bile acid intended for research and analytical applications. Typical pack size is 50 MG; chemical formula C26H43NO6 and molecular weight ~465.6 g/mol. Purity is reported as ≥98% by some suppliers; verify the certificate of analysis.

  • Analytical reference standard for use in research and analytical assays.
  • Glycine-conjugated bile acid structure, useful as a metabolite standard.
  • Chemical formula: C26H43NO6.
  • Molecular weight: ~465.6 g/mol.
  • Supplied as a 50 MG solid suitable for analytical preparation.
  • Purity reported as ≥98% by some suppliers; check the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.