Learn More
Cayman Chemical Glycohyocholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502938042
Encompass_Preferred
An internal standard for the quantification of glycohyocholic acid by GC- or LC-MS
Catalog No. 50-293-8042 Supplier Cayman Chemical Supplier No. 270341 MG
An internal standard for the quantification of glycohyocholic acid by GC- or LC-MS