Learn More
Cayman Chemical GlycohyodeoxycholIc AcId 10mg
SDP
Catalog No. 502995576
Encompass_Preferred
A major metabolite of hyodeoxycholic acid in humans; supplementation in prairie dogs fed a lithogenic diet decreases the frequency of cholesterol crystals in the gallbladder; inhibits the activity of cholesterol 7α-hydroxylase
Catalog No. 50-299-5576 Supplier Cayman Chemical Supplier No. 2264310 MG
A major metabolite of hyodeoxycholic acid in humans; supplementation in prairie dogs fed a lithogenic diet decreases the frequency of cholesterol crystals in the gallbladder; inhibits the activity of cholesterol 7α-hydroxylase