Learn More
Cayman Chemical Glycohyodeoxycholic Acid, Purity: ≥95%, CAS: 13042-33-6, 50 mg
SDP
Catalog No. 502994122
Encompass_Preferred
A major metabolite of hyodeoxycholic acid in humans; supplementation in prairie dogs fed a lithogenic diet decreases the frequency of cholesterol crystals in the gallbladder; inhibits the activity of cholesterol 7α-hydroxylase
Catalog No. 50-299-4122 Supplier Cayman Chemical Supplier No. 2264350 MG
A major metabolite of hyodeoxycholic acid in humans; supplementation in prairie dogs fed a lithogenic diet decreases the frequency of cholesterol crystals in the gallbladder; inhibits the activity of cholesterol 7α-hydroxylase