Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Glycol/Formaldehyde Refill Kit
SDP

Catalog No. 13299620
Catalog No. 13-299-620 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R4815
Add to Cart
Add to Cart

Refill kit for glycol and formaldehyde visual test kits. Contains 30 self-filling ampoules packaged securely in trays and a box.

  • Periodic acid oxidizes ethylene glycol and propylene glycol to formaldehyde.
  • In a highly alkaline solution, and in conjunction with the oxidizing agent persulfate, formaldehyde reacts with Purpald to form a purple-colored complex. Test results are expressed as ppm (mg/L) ethylene glycol. To convert results to ppm propylene glycol, multiply by 2. Note: This chemistry does not differentiate between ethylene and propylene glycol.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-4815 or K-4605
No. of Tests 30
Type Glycol/Formaldehyde Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.