Learn More
Medchemexpress LLC Glycol salicylate | 87-28-5 | 98.8% | 1 G
SDP
Catalog No. 5000411435
Encompass_Preferred
Glycol salicylate is a derivative of salicylic acid that effectively moisturizes the stratum corneum and improves the aesthetic appearance of the skin. It is hygroscopic, and its hydrated solution exhibits a human epidermal flux 10 times higher than the pure solution. This results in a higher diffusion rate in the stratum corneum and a physiologically induced self-enhancement effect in absorption in vivo.
  • Effectively moisturizes the stratum corneum.
  • Improves aesthetic appearance of the skin.
  • Hygroscopic with high epidermal flux in hydrated solution.
  • Increased diffusion rate in stratum corneum.
  • Physiologically induced self-enhancement effect in absorption in vivo.
Catalog No. 50-004-11435 Supplier Medchemexpress LLC Supplier No. HYB22081G
Glycol salicylate is a derivative of salicylic acid that effectively moisturizes the stratum corneum and improves the aesthetic appearance of the skin. It is hygroscopic, and its hydrated solution exhibits a human epidermal flux 10 times higher than the pure solution. This results in a higher diffusion rate in the stratum corneum and a physiologically induced self-enhancement effect in absorption in vivo.
  • Effectively moisturizes the stratum corneum.
  • Improves aesthetic appearance of the skin.
  • Hygroscopic with high epidermal flux in hydrated solution.
  • Increased diffusion rate in stratum corneum.
  • Physiologically induced self-enhancement effect in absorption in vivo.