Learn More
Hach Company Glycol test Reagent 2 Powder Pillows, pk/25
Supplier:  Hach Company 2183768
Encompass
For presence/absence detection of glycols by visual test kit. Replacement reagent for Hach Glycol Detection Test Kit - 2186400. Pack of 25 Powder Pillows.
Catalog No. 50-199-5520