Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolaldehyde-1-13C | 71122-42-4 | 99.0% | C13CH4O2 | 10MG
SDP

Catalog No. 5000258390
Encompass_Preferred

Glycolaldehyde-1-13C is the 13C-labeled form of glycolaldehyde for research use as a stable-isotope tracer and analytical standard. It is supplied as a high-purity aqueous solution suitable for mass spectrometry and NMR applications.

  • Isotopic label at carbon 1 (13C).
  • High chemical purity: 99.0%.
  • High isotopic enrichment (reported ≥99% atom 13C).
  • Appearance: colorless to light yellow aqueous solution.
  • Intended uses include tracer studies, analytical standards, and NMR reference materials.
  • Common research sizes available: 1 mg-100 mg.

Catalog No. 50-002-58390 Supplier Medchemexpress LLC Supplier No. HY146995S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycolaldehyde-1-13C is the 13C-labeled form of glycolaldehyde for research use as a stable-isotope tracer and analytical standard. It is supplied as a high-purity aqueous solution suitable for mass spectrometry and NMR applications.

  • Isotopic label at carbon 1 (13C).
  • High chemical purity: 99.0%.
  • High isotopic enrichment (reported ≥99% atom 13C).
  • Appearance: colorless to light yellow aqueous solution.
  • Intended uses include tracer studies, analytical standards, and NMR reference materials.
  • Common research sizes available: 1 mg-100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.