Learn More
Ethos Biosciences Glycolic Acid 20PCT 2oz
Catalog No. 5000819204
Encompass_Preferred
Glycolic Acid 20% is a concentrated aqueous alpha hydroxy acid (AHA) solution suitable for general laboratory cosmetic formulation and research applications Commonly used as a chemical reagent and formulation component it provides consistent concentration for reliable and reproducible results The solution is appropriate for product development pH adjustment and general chemical processing tasks Supplied in chemically compatible containers Glycolic Acid 20% is intended for professional laboratory and industrial use Not intended for direct consumer use without appropriate formulation and regulatory review
Catalog No. 50-008-19204 Supplier Ethos Biosciences Supplier No. 354902
Glycolic Acid 20% is a concentrated aqueous alpha hydroxy acid (AHA) solution suitable for general laboratory cosmetic formulation and research applications Commonly used as a chemical reagent and formulation component it provides consistent concentration for reliable and reproducible results The solution is appropriate for product development pH adjustment and general chemical processing tasks Supplied in chemically compatible containers Glycolic Acid 20% is intended for professional laboratory and industrial use Not intended for direct consumer use without appropriate formulation and regulatory review