Learn More
Medchemexpress LLC Glycolic acid | 79-14-1 | MFCD00004312 | 97.0% | 76.05 g/mol | C2H4O3 | 1 ML
SDP
Catalog No. 5000303900
Encompass_Preferred
Glycolic acid (hydroxyethanoic acid) is a small organic acid used in research to study pigmentation and biochemical processes. Supplied as a 10 mM solution in DMSO for research and analytical applications, it acts as a tyrosinase inhibitor that can suppress melanin formation and is commonly used in studies of skin-lightening mechanisms and topical formulation research. Purity is typically reported at about 97.0%.
  • Tyrosinase inhibitor for studies of melanin formation.
  • Ready-to-use 10 mM solution in DMSO for convenience.
  • Suitable for research and analytical applications.
  • High purity (approximately 97.0%) for reproducible results.
  • Low molecular weight and water-soluble moiety support biochemical assays.
Catalog No. 50-003-03900 Supplier Medchemexpress LLC Supplier No. HYW01596710MM/1ML
Glycolic acid (hydroxyethanoic acid) is a small organic acid used in research to study pigmentation and biochemical processes. Supplied as a 10 mM solution in DMSO for research and analytical applications, it acts as a tyrosinase inhibitor that can suppress melanin formation and is commonly used in studies of skin-lightening mechanisms and topical formulation research. Purity is typically reported at about 97.0%.
  • Tyrosinase inhibitor for studies of melanin formation.
  • Ready-to-use 10 mM solution in DMSO for convenience.
  • Suitable for research and analytical applications.
  • High purity (approximately 97.0%) for reproducible results.
  • Low molecular weight and water-soluble moiety support biochemical assays.