Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolic acid | 79-14-1 | MFCD00004312 | ≥97.0% | 76.05 g/mol | C2H4O3 | 500 MG
SDP

Catalog No. 5000301418
Encompass_Preferred

Glycolic acid is a small, water-soluble alpha-hydroxy acid used as an analytical reagent and biochemical standard. It acts as a tyrosinase inhibitor and is supplied as a crystalline solid for research and analytical applications; safety, assay, and storage information are provided in the SDS and COA.

  • Purity ≥97.0%.
  • Molecular weight 76.05 g/mol.
  • Chemical formula C2H4O3.
  • White to off-white solid.
  • Intended for research and analytical use.
  • Causes severe skin burns and eye damage; follow SDS handling instructions.
  • Store at room temperature; stability data available in COA.

Catalog No. 50-003-01418 Supplier Medchemexpress LLC Supplier No. HYW015967500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycolic acid is a small, water-soluble alpha-hydroxy acid used as an analytical reagent and biochemical standard. It acts as a tyrosinase inhibitor and is supplied as a crystalline solid for research and analytical applications; safety, assay, and storage information are provided in the SDS and COA.

  • Purity ≥97.0%.
  • Molecular weight 76.05 g/mol.
  • Chemical formula C2H4O3.
  • White to off-white solid.
  • Intended for research and analytical use.
  • Causes severe skin burns and eye damage; follow SDS handling instructions.
  • Store at room temperature; stability data available in COA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.