Learn More
Medchemexpress LLC Glycolic acid-d2 | 75502-10-2 | 98.5% | 78.06 | C2H2D2O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW015967S1MG
Encompass_Preferred
Glycolic acid-d2 is the deuterium-labeled analogue of glycolic acid intended for research and analytical applications. It is commonly used as an internal standard and tracer in mass spectrometry and isotopic labeling studies, and supports investigations of tyrosinase inhibition and melanin biosynthesis.
  • Deuterium-labeled analogue for isotope tracing and quantitation.
  • Suitable as an internal standard for mass spectrometry and analytical workflows.
  • High purity (reported ~98.5%) for reliable analytical results.
  • Solid, white to off-white form for easy handling and storage.
  • Available in small pack sizes for trace-level experiments.
  • Recommended storage conditions to preserve compound integrity.
Catalog No. 50-003-00577