Learn More
Medchemexpress LLC Glycolic acid-d2 | 75502-10-2 | 98.5% | 78.06 | C2H2D2O3 | 5 MG
SDP
Catalog No. 5000303898
Encompass_Preferred
Glycolic acid-d2 is the deuterium-labeled analogue of glycolic acid used as an analytical standard and tracer in biochemical and cosmetic research. It functions as a tyrosinase inhibitor that suppresses melanin formation and is suitable for metabolic, enzymatic, and mass-spectrometry studies requiring isotopic labeling.
  • Deuterium-labeled for mass spectrometry tracing and isotope studies.
  • Reported high purity suitable for analytical applications.
  • White to off-white solid form for easy handling and storage.
  • Stable when stored frozen; recommended conditions available in product documentation.
  • Available in small packaging sizes for use as standards and internal controls.
Catalog No. 50-003-03898 Supplier Medchemexpress LLC Supplier No. HYW015967S5MG
Glycolic acid-d2 is the deuterium-labeled analogue of glycolic acid used as an analytical standard and tracer in biochemical and cosmetic research. It functions as a tyrosinase inhibitor that suppresses melanin formation and is suitable for metabolic, enzymatic, and mass-spectrometry studies requiring isotopic labeling.
  • Deuterium-labeled for mass spectrometry tracing and isotope studies.
  • Reported high purity suitable for analytical applications.
  • White to off-white solid form for easy handling and storage.
  • Stable when stored frozen; recommended conditions available in product documentation.
  • Available in small packaging sizes for use as standards and internal controls.