Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolic acid-d2 | 75502-10-2 | MFCD00209729 | 98.5% | 78.06 | C2H2D2O3 | 10 MG
SDP

Catalog No. 5000303887
Encompass_Preferred

Glycolic acid-d2 is the deuterium-labeled form of glycolic acid for research and analytical applications. It can be used as a stable isotope standard or tracer and shares glycolic acid's reported tyrosinase inhibitory activity; supplied for laboratory use only.

  • Deuterium-labeled reagent for tracer and mass spectrometry applications.
  • High chemical purity: 98.5%.
  • Molecular formula C2H2D2O3 and molecular weight 78.06.
  • Available in small pack sizes suitable for analytical workflows (1, 5, 10 mg; larger sizes on request).
  • Storage-stable as powder: -20°C (3 years) or 4°C (2 years).
  • Supplied with datasheet, certificate of analysis, and safety data sheet for quality control.

Catalog No. 50-003-03887 Supplier Medchemexpress LLC Supplier No. HYW015967S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycolic acid-d2 is the deuterium-labeled form of glycolic acid for research and analytical applications. It can be used as a stable isotope standard or tracer and shares glycolic acid's reported tyrosinase inhibitory activity; supplied for laboratory use only.

  • Deuterium-labeled reagent for tracer and mass spectrometry applications.
  • High chemical purity: 98.5%.
  • Molecular formula C2H2D2O3 and molecular weight 78.06.
  • Available in small pack sizes suitable for analytical workflows (1, 5, 10 mg; larger sizes on request).
  • Storage-stable as powder: -20°C (3 years) or 4°C (2 years).
  • Supplied with datasheet, certificate of analysis, and safety data sheet for quality control.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.