Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycolic acid ethoxylate oleyl ether

Catalog No. 463280100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-328-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-328-0100ML Supplier Sigma Aldrich Supplier No. 463280100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Anionic surfactant. Emulsifier for metal-working fluids.

Specifications

CAS 57635-48-0
Density 0.99 g/mL (at 25°C)
Molecular Formula C55H112O8
Refractive Index n20/D 1.461
Linear Formula CH3(CH2)xCH=CH(CH2)8(OCH2CH2)yOCH2CO2H, x = 5-7, y ~2
Quantity 100 mL
MDL Number MFCD00679048
Synonym Oleth-10 carboxylic acid; Oleth-3 carboxylic acid; Oleth-6 carboxylic acid
Molecular Weight (g/mol) 901.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.