Learn More
Medchemexpress LLC Glycolic acid oxidase inhibitor 1 | 77529-42-1 | 99.4% | 344.16 | 1 ML
SDP
Catalog No. 5000400757
Encompass_Preferred
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1. It inhibits oxalate biosynthesis in isolated perfused rat liver. Glycolate oxidase inhibitors provide a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.
  • Glycolate oxidase inhibitor
  • Inhibits oxalate biosynthesis
  • Potential for prevention and treatment of calcium oxalate renal lithiasis
Catalog No. 50-004-00757 Supplier Medchemexpress LLC Supplier No. HY2216610MM/1ML
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1. It inhibits oxalate biosynthesis in isolated perfused rat liver. Glycolate oxidase inhibitors provide a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.
  • Glycolate oxidase inhibitor
  • Inhibits oxalate biosynthesis
  • Potential for prevention and treatment of calcium oxalate renal lithiasis